Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate, Anhydrous, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1062689012 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
12 kg
25 kg
Packaging:
Fiber Carton
Plastic Bottle
Plastic Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1062689012 12 kg Plastic Drum
M1062680250 250 g Plastic Bottle
M1062681000 1 kg Plastic Bottle
M1062682500 2.5 kg Plastic Bottle
M1062689025 25 kg Fiber Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. M1062689012 Supplier MilliporeSigma Supplier No. 1062689012
Add to Cart
Add to Cart

Recommended Storage

+5°C to +30°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Specifications

Boiling Point >400°C (decomposition)
Melting Point 324°C (decomposition)
Color Colorless
pH 7.5 to 9.2 (30g/L, H2O, 20°C)
Physical Form Powder
Quantity 12 kg
Solubility Information 365g/L
Formula Weight 82.03
Percent Purity >99.0%
Grade ACS
Packaging Plastic Drum
Density 1.52 g/3 (20 °C)
Chemical Name or Material Sodium Acetate
Show More Show Less
EINECSNumber 204-823-8
RTECSNumber AJ4580000
Recommended Storage +5°C to +30°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.