Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate Trihydrate, ACS Reagent, Reagents

Catalog No. 01337762
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Catalog No. Quantity
01-337-762 2.5 kg
01-337-764 100 g
01-337-763 10 kg
01-337-761 500 g
4 options

Catalog No. 01-337-762

Supplier: Reagents Holdings Llc C13065002.5A5

Only null left
Add to Cart
Add to Cart

Sodium Acetate Trihydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.08
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
IUPAC Name sodium acetate trihydrate
SMILES O.O.O.[Na+].CC([O-])=O

Specifications

Boiling Point 123°C
Color Colorless/White
Physical Form Solid
Quantity 2.5 kg
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.45
Chemical Name or Material Sodium Acetate Trihydrate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.