Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Acetate, Trihydrate, Crystal, Ultrapure Bioreagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
Description
- For Liquid Chromatography and Molecular Biology applications
Specifications
Specifications
Physical Form | Crystals |
Quantity | 1 kg |
Formula Weight | 136.08 |
Grade | Ultrapure Bioreagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.45kg |
Chemical Name or Material | Sodium Acetate, Trihydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction