Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate Trihydrate (White Crystals/USP/FCC/EP/BP), Fisher Chemical™

Catalog No. p-33590 Shop All Fisherbrand Products
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
MDL Number MFCD00071557
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Melting Point 58°C
Color White
pH 8 to 9.5
Physical Form Solid
Quantity 2.5 kg
Linear Formula CH3COONa·3H2O
Formula Weight 136.08
Percent Purity 99.0 to 101.0% (FCC)
Grade BP/EP/FCC/USP
Identification Pass Test (BP), Pass Test (EP), Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC/EP/BP)
Loss on Drying 36.0 to 41.0% (FCC), 36.0 to 41.0% (USP/FCC), 39.0 to 40.5% (EP/BP), 38.0 to 41.0% (USP), 39.0 to 40.5% (BP), 39.0 to 40.5% (EP)
Packaging Poly Bottle
Density 1.45g/cm³
Chemical Name or Material Sodium Acetate Trihydrate
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.