Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Alginate, FCC, 90.8-106%, Spectrum™

Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
9005-38-3
(C6H7O7)A(C6H7O7)BNa
NaN
[Na+].O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C([O-])=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C([O-])=O
9005-38-3
(C6H7O7)A(C6H7O7)BNa
NaN
[Na+].O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C([O-])=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C([O-])=O

SO106, 9005-38-3, (C6H7O6Na)n

Spectrum™ Sodium Alginate, FCC is a gum that has no discernible flavor and is used in the food industry as an emulsifier and to increase viscosity. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Arsenic (As) 3 mg/kg
Percent Purity 90.8 to 106%
Lead (Pb) 5 mg/kg
Grade FCC
Loss on Drying 0.15
Packaging Fiber Drum
Quantity 50 kg