Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium aluminum hydride, 93%, Thermo Scientific Chemicals

Catalog No. AC380115000
Click to view available options
Quantity:
100 g
500 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13770-96-2
Molecular Formula H4AlNa
Molecular Weight (g/mol) 54
MDL Number MFCD00192443
InChI Key MNYCIYRTKITUKR-UHFFFAOYSA-N
Synonym sodium alumanuide, sodium aluminum hydride, aluminum sodium hydride, aluminumsodiumhydride, naalh4, sah22, sodiumtetrahydroaluminate, sodiumaluminumtetrahydride, na alh4, sodium tetrahydroaluminate
PubChem CID 26266
ChEBI CHEBI:30141
IUPAC Name sodium;alumanuide
SMILES [Na+].[AlH4-]

Specifications

Melting Point 178.0°C
Quantity 500 g
Assay Percent Range 93%
Solubility Information Solubility in water: reacts violently
Formula Weight 54
Percent Purity 93%
Chemical Name or Material Sodium Aluminum hydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.