Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium amide, 95%, pure

Catalog No. AC197050010
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC197050010 1 kg Glass bottle
AC197051000 100 g Glass bottle
AC197055000 500 g Glass bottle
AC197050025 2.5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC197050010 Supplier Thermo Scientific Chemicals Supplier No. 197050010

CAS: 7782-92-5 | H2NNa | 39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-92-5
InChI Key ODZPKZBBUMBTMG-UHFFFAOYSA-N
Synonym sodium amide, sodamide, sodium amide na nh2, unii-5db3g6px9d, 5db3g6px9d, sodium azanide, sodium amide, 50 wt. % suspension in toluene, sodiumamide, natriumamid, sodioamine
PubChem CID 24533
IUPAC Name sodium;azanide
SMILES [NH2-].[Na+]

Specifications

Color Gray-White to White
Physical Form Powder, Irregular Flakes or Granules
Quantity 1 kg
Assay Percent Range 95%
Molecular Formula H2NNa
Linear Formula NaNH2
Fieser 01,1034; 02,373; 04,439; 05,591; 06,525; 08,446; 15,288
Molecular Weight (g/mol) 39.013
Formula Weight 39
Percent Purity ≥94% (total base)
Grade Pure
Packaging Glass bottle
Chemical Name or Material Sodium amide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.