Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium aurothiomalate(I), 99.9% (metals basis)

Catalog No. p-7050930
Click to view available options
Quantity:
1 g
5 g

CAS: 12244-57-4 | C4H5AuNa2O5S | 408.09 g/mol

Sodium aurothiomalate(I) is useful for high mobility group box chromosomal protein 1 western blotting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12244-57-4
Molecular Formula C4H5AuNa2O5S
Molecular Weight (g/mol) 408.09
MDL Number MFCD00064304,MFCD00064304
InChI Key YLQOAPBVYJCTPW-UHFFFAOYNA-K
Synonym gold sodium thiomalate
PubChem CID 133108869
IUPAC Name gold;sodium;2-sulfanylbutanedioic acid
SMILES O.[Na+].[Na+].[Au+].[O-]C(=O)CC([S-])C([O-])=O

Specifications

Odor Sulphurous
Quantity 1 g
Merck Index 14,4518
Solubility Information Very soluble in water; Practically insoluble in alcohol, ether
Formula Weight 390.08
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Sodium aurothiomalate(I)
Hazard Category H302+H332-H317
Hazard Statement GHS H Statement
H301-H332-H317
Toxic if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280g-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P363-P501c
EINECSNumber 235-479-7
RTECSNumber MD5435000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.