Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bicarbonate, 1 M Buffer Solution, pH 10.0, PROMO, Thermo Scientific Chemicals

Catalog No. J63025APPM
Click to view available options
Quantity:
250 mL
500 mL

Applications

The bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.

Solubility

It is soluble in water.

Notes

Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents and strong acids.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01 g/mol
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Concentration 1 M
pH 10.0
Physical Form Liquid
Quantity 500 mL
Solubility Information It is soluble in water.
Chemical Name or Material Sodium bicarbonate buffer solution

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.