Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bicarbonate (Crystalline Powder/USP/FCC/EP/BP/JP), Fisher Chemical™

Catalog No. p-46823 Shop All Fisherbrand Products
Click to view available options
Quantity:
12 kg
Packaging:
Poly Pail

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Melting Point 270°C
Color White
pH 8.3
Physical Form Powder/Solid
Quantity 12 kg
Solubility Information Pass Test
Formula Weight 84.01
Percent Purity 99.0 to 101.0% (EP)
Grade BP/EP/FCC/JP/USP
Identification Pass Test (BP), Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (USP), Pass Test (USP/FCC/EP/BP/JP)
Loss on Drying 0.25% max. (FCC), 0.25% max. (USP), 0.25% max. (USP, FCC)
Packaging Poly Pail
Decomposition Information >50°C
Chemical Name or Material Sodium Bicarbonate
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes and skin. May cause adverse kidney effects. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.