Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bicarbonate, MP Biomedicals™

Catalog No. MP021914355 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021914355 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021914355 Supplier MP Biomedicals, Inc Supplier No. 02191435.5
Add to Cart
Add to Cart

Can be used as a neutralizer, since it reacts with both acids and bases. Sodium Bicarbonate, MP Biomedicals is a commonly used laboratory chemical, also referred to as baking soda. It can be an effective way of controlling fungus growth or to remove impur

When reacting with bases, the compound forms carbonates. When reacting with carboxyl groups, it gives a fast-forming and effervescent carbon dioxide formation. Its many uses include:

  • Remove or washes acidic impurities from a crude liquid to yield a purer sample
  • To test for the presence of carboxylic groups in proteins
  • Decreases serum potassium levels, as well as increases blood pH and bicarbonate concentration
  • Decreases the duration of a peripheral nerve block in rats when added to Lidocaine
  • Effective way of controlling fungus growth
  • When mixed with water, can be used as an antacid
  • Used as a leavening agent in cooking and baking (CO2 release causes expansion of the batter and forms the characteristic texture and grain in baked and fried foods)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 84.01
Percent Purity 99.7 to 100.3%
Grade ACS Reagent
Recommended Storage Room Temperature (15-30°C)
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.