Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bicarbonate, Powder, USP, EP, BP, JP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606339 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
2.5 kg
6 x 500 g
12 kg
25 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

Catalog No. 18-606-339

Supplier: Spectrum Chemical Manufacturing Corporation SO12525KGBL

Only null left
Add to Cart
Add to Cart

SO125, 144-55-8, NaHCO3

Spectrum™ Chemical Sodium Bicarbonate, Powder, USP, EP, BP, JP is used as an antacid to treat heartburn, indigestion, and upset stomach. All Spectrum Chemical USP, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

pH 7.9 to 8.4
CAS Max % 1
Quantity 25 kg
Percent Purity 99 to 100.5%
Grade Multi-Compendial
Loss on Drying 0.0025
Packaging Fiber Drum
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.