Learn More
Sodium bismuth oxide, 80%
Description
Sodium bismuthate oxide is used as an oxidant in the preparation of hexavalent Americium (Am) during the separation of Am from the lanthanides., hydrothermal synthesis of bismuth oxychloride (BiOCl) micrometer-sized single crystals. It is also used as a precursor in the synthesis of bismuth nanoforms under microwave (MW) irradiation. It used to converts almost any manganese compound to permanganate, which is easily assayed spectrophotometrically.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium bismuthate oxide is used as an oxidant in the preparation of hexavalent Americium (Am) during the separation of Am from the lanthanides., hydrothermal synthesis of bismuth oxychloride (BiOCl) micrometer-sized single crystals. It is also used as a precursor in the synthesis of bismuth nanoforms under microwave (MW) irradiation. It used to converts almost any manganese compound to permanganate, which is easily assayed spectrophotometrically.
Solubility
Soluble in aqueous acid. Insoluble in water.
Notes
Hygroscopic. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.

Specifications
Specifications
Quantity | 100 g |
Merck Index | 14,8585 |
Solubility Information | Soluble in aqueous acid. Insoluble in water. |
Formula Weight | 279.97 |
Percent Purity | 80% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Chemical Name or Material | Sodium bismuth oxide |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 235-455-6 |
RTECSNumber | VZ1750000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.