Learn More
Sodium bismuth oxide, ACS, 80% min
Description
Sodium bismuth oxide is used for the determination of manganese in iron, steel and ores. It is also used in the separation of plutonium and acts as a strong oxidizer. It is used in the conversion of manganese compound to permanganate. Further, it is used in the preparation of hexavalent americium (Am) during its separation from the lanthanides and in the hydrothermal synthesis of bismuth oxychloride micrometer-sized single crystals. In addition to this, it is employed as a precursor in the preparation of bismuth nanoforms.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Yellow to Brown |
Physical Form | Powder |
Quantity | 25 g |
Merck Index | 14,8585 |
Solubility Information | Insoluble in cold water; Decomposes in hot water forming Bi2O3,NaOH,and oxygenInsoluble in water. |
Formula Weight | 279.97 |
Percent Purity | ≤80% |
Grade | ACS |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
missing translation for 'healthHazard1' | H302-H315-H319-H335 |
missing translation for 'healthHazard2' | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
missing translation for 'healthHazard3' | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 235-455-6 |
RTECSNumber | VZ1750000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.