Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium bismuthate(V), 85%
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow to Brown |
| Physical Form | Powder |
| Quantity | 250 g |
| Assay Percent Range | 80% min. (Iodometry) |
| Linear Formula | NaBiO3 |
| Merck Index | 15,8721 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: decomposes in hot water and acids |
| Formula Weight | 279.96 |
| Percent Purity | 85% |
| Packaging | Glass bottle |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wash face,hands and any exposed skin thoroughly after handling. Do not eat,drink or smoke when using this product. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. If skin irritation occurs: Get medical advice/a |
| EINECSNumber | 235-455-6 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction