Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bisulfate, Monohydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

10034-88-5
H3NaO5S
138.069
JXHZRQHZVYDRGX-UHFFFAOYSA-M
23673662

High quality chemicals for laboratory and specialized industrial use

SMILES O.OS(=O)(=O)[O-].[Na+]
IUPAC Name sodium;hydrogen sulfate;hydrate
Formula Weight 138.07
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.10kg
Quantity 500g
Chemical Name or Material Sodium Bisulfate, Monohydrate
Grade BAKER ANALYZED™ Reagent