Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bitartrate, Monohydrate, Powder, Reagent, 99%, Spectrum™ Chemical
SDP

Catalog No. 18606349
Click to view available options
Quantity:
125 g
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle

S1175, 6131-98-2, NaHC4H4O6·H2O

Spectrum™ Chemical Sodium Bitartrate, Monohydrate, Powder, Reagent The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 6131-98-2
Molecular Formula C4H7NaO7
Molecular Weight (g/mol) 190.08
InChI Key LLVQEXSQFBTIRD-UHFFFAOYNA-M
IUPAC Name sodium 3-carboxy-2,3-dihydroxypropanoate hydrate
SMILES O.[Na+].OC(C(O)C([O-])=O)C(O)=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 99%
Grade Reagent
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.