Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borate, Decahydrate, Crystal, Reagent, ACS, 99.5-105%, Spectrum™ Chemical
SDP

Catalog No. p-7158189
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

S1180, 1303-96-4, Na2B4O7·10H2O

Spectrum™ Chemical Sodium Borate, Decahydrate, Crystal, Reagent, ACS is also known simply as Borax and is a mineral that occurs naturally in deposits of the mineral Evaporite. Its uses vary from household products, such as hand soap and water softening agents, to use as a flux in welding and can also be found in small scale gold mining operations as a substitute to mercury. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4H20Na2O17
Molecular Weight (g/mol) 381.36
InChI Key OJPFLODMGUZMKD-UHFFFAOYSA-N
IUPAC Name disodium decahydrate oxoboranyl ({oxido[(oxoboranyl)oxy]boranyl}oxy)boronate
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B(OB=O)OB([O-])OB=O

Specifications

pH 9.15 to 9.20
CAS Max % 1
Quantity 500 g
Percent Purity 99.5 to 105%
Grade ACS
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.