Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium borodeuteride, 99% (Isotopic)

Catalog No. p-7050061
Click to view available options
Quantity:
1 g
5 g

CAS: 15681-89-7 | BH4Na | 37.83 g/mol

Sodium borodeuteride is used to prepare alfa, alfa-dideutero-beta-amino acids. It is also used to prepare amine-stabilized, deuterated borane-tetrhydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15681-89-7
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003519
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
Synonym sodium borodeuteride, sodium tetradeuterioboron, sodium borohydride-d4, sodium 2h4 tetrahydroborate 1-, sulfatase, borate 1-, tetrahydro-d4-, sodium, nabd4, sodium 2h4 boranuide, deuterated sodium borohydride
PubChem CID 23673181
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Physical Form Powder
Color White
Sensitivity Moisture
Quantity 1 g
pH 11
Melting Point ∼400°C (decomposition)
Linear Formula NaBD4
UN Number UN1426
Solubility Information Soluble in liquid ammonia,amines and pyridine.
Formula Weight 41.88
Percent Purity 99%
Chemical Name or Material Sodium borodeuteride, Isotopic
Show More Show Less
Hazard Category H261-H301-H314
Hazard Statement GHS H Statement
H301-H311-H330-H314-H318-H261
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
In contact with water releases flammable gas.
Precautionary Statement P223-P231+P232-P260-P264b-P270-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P378q-P380-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: SODIUM BOROHYDRIDE
EINECSNumber 239-764-7
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.