Sodium Borohydride, Caplets, 98%, Alfa Aesar™

Manufacturer: Alfa Aesar 3878822

Quantity 100g

 View more versions of this product

Catalog No. AA3878822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16940-66-2
    Molecular Formula NaBH4
    Formula Weight 37.83
    MDL Number MFCD00003518
    Synonym Sodium tetrahydridoborate
    Chemical Name or Material Sodium Borohydride

    Specifications

    Physical Form Caplets
    Formula Weight 37.83
    Color White
    Quantity 100g
    Density 1.074g/cm3 at 20°C
    EINECS Number 241-004-4
    Assay Percent Range 98%
    MDL Number MFCD00003518
    CAS 16940-66-2
    Synonym Sodium tetrahydridoborate
    Ignition Point 220°C (428°F)
    Odor Odorless
    Molecular Formula NaBH4
    Appearance White caplets
    Health Hazard 1 H301-H311-H331-H314-H318-H261: Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. In contact with water releases flammable gas.
    Chemical Name or Material Sodium Borohydride

    H301-H311-H331-H314-H318-H261: Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. In contact with water releases flammable gas.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit