Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium borohydride, 99%, (VenPureTM SF granules), granules

Catalog No. p-6827186
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum

CAS: 16940-66-2 | BH4Na | 37.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
Synonym Sodium tetrahydroborate, SBH
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Melting Point 360.0°C
Quantity 100 g
Assay Percent Range 98.5% min. (Hydrogen evolution)
Linear Formula NaBH4
Merck Index 15,8727
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents
Formula Weight 37.83
Percent Purity 99%
Packaging Plastic Bottle
Chemical Name or Material Sodium borohydride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
May damage fertility.
May damage the unborn child.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts viol
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
IF INHALED: Remove to fresh air and keep at rest in a position comf
EINECSNumber 241-004-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.