Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Borohydride, Crystalline Powder, Reagent, ACS, 98%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
40 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
16940-66-2
BH4Na
37.83
YOQDYZUWIQVZSF-UHFFFAOYSA-N
sodium boranuide
[BH4-].[Na+]
16940-66-2
BH4Na
37.83
YOQDYZUWIQVZSF-UHFFFAOYSA-N
sodium boranuide
[BH4-].[Na+]

S1187, 16940-66-2, NaBH4

Spectrum™ Sodium Borohydride, Crystalline Powder, Reagent, ACS is an inorganic compound that is white in color and usually found as a powder. It is a reducing agent with versatility and is used as a synthetic reagent in the laboratory. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Grade ACS
CAS Max % 1
Percent Purity 98%
Packaging Fiber Drum
Quantity 40 kg