Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bromide, ≥99.0%, ACS Reagent, Solstice

Catalog No. 60045887 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-887 100 g Poly Bottle
60-045-888 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-887 Supplier Solstice Supplier No. 310506100G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
IUPAC Name sodium;bromide
SMILES [Na+].[Br-]

Specifications

Boiling Point 1393°C
Melting Point 755°C
Quantity 100 g
Linear Formula NaBr
UN Number NONH for all modes of transport
Solubility Information Soluble in water and ethanol
Formula Weight 102.89g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Vapor Pressure 1mmHg (806°C)
Density 3.2g/cm3
Chemical Name or Material Sodium Bromide
Show More Show Less
EINECSNumber 231-599-9
RTECSNumber VZ3150000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.