Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bromide, Puratronic™, 99.9955% (metals basis)

Catalog No. p-4407865
Click to view available options
Quantity:
25 g
100 g

CAS: 7647-15-6 | BrNa | 102.89 g/mol

Sodium bromide is widely used for the preparation of other bromides in organic synthesis and other areas. Also used to prepare dense fluids used in oil wells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390°C
Melting Point 755°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9955% (metals basis)
Solubility Information Soluble in water. Also soluble in alcohol,methanol and ethanol.
IUPAC Name sodium bromide
Formula Weight 102.89
Refractive Index 1.6412
Sensitivity Hygroscopic
Appearance Powder/flake/crystalline/beads
Density 3.203 g/cm3 at 20°C
Chemical Name or Material Sodium bromide
Show More Show Less
EINECSNumber 231-599-9
RTECSNumber VZ3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.