Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Cacodylate, 0.1 M Buffer Solution, pH 7.0, PROMO, Thermo Scientific™

Catalog No. J60344AEPM
Click to view available options
Quantity:
100 mL

Chemical Identifiers

CAS 6131-99-3
Molecular Formula C2H6AsNaO2
Molecular Weight (g/mol) 159.98 g/mol
MDL Number MFCD00149079
InChI Key IHQKEDIOMGYHEB-UHFFFAOYSA-M
IUPAC Name sodium dimethylarsinate
SMILES [Na+].C[As](C)([O-])=O

Specifications

Physical Form Liquid
Quantity 100 mL
ShelfLife 5 Years

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.