Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Sodium cacodylate, 0.1M buffer soln., pH 6.8
Click to view available options
Quantity:
50 mL
Specifications
Specifications
CAS | 6131-99-3 |
Physical Form | Liquid |
Quantity | 50 mL |
MDL Number | MFCD00149079 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction