Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium caprylate, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
1984-06-1
C8H15NaO2
166.2
MFCD00058511
BYKRNSHANADUFY-UHFFFAOYSA-M
sodium octanoate,sodium caprylate,octanoic acid, sodium salt,sodium n-octanoate,sodium octoate,natrium octanoat,caprylic acid sodium salt,octanoic acid sodium salt,octanoic acid, sodium salt 1:1,unii-9xtm81vk2b
23664772
1984-06-1
C8H15NaO2
166.2
MFCD00058511
BYKRNSHANADUFY-UHFFFAOYSA-M
sodium octanoate,sodium caprylate,octanoic acid, sodium salt,sodium n-octanoate,sodium octoate,natrium octanoat,caprylic acid sodium salt,octanoic acid sodium salt,octanoic acid, sodium salt 1:1,unii-9xtm81vk2b
23664772
Melting Point >225°C
Quantity 500g
Color White
Infrared Spectrum Authentic
Packaging Plastic bottle
Water 5% max. (K.F.)
Assay Percent Range 98%
Solubility Information Solubility in water: soluble in water
Formula Weight 166.2
Physical Form Powder
Percent Purity ≥97.5% (on dry substance)
Chemical Name or Material Sodium caprylate, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 217-850-5

RTECSNumber : RH0787000

TSCA : TSCA