Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, 0.0454 N, 1 mL = 1 mg CO2, Ricca Chemical
SDP

Catalog No. p-7239413 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L

APHA for Carbon Dioxide Testing

Specifications

Chemical Name or Material Sodium Carbonate
Color Colorless
CAS 497-19-8, 7732-18-5
CAS Min % 98.0, 0.24
CAS Max % 100.0, 0.25
Molecular Formula CNa2O3
MDL Number MFCD00003494
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C([O-])=O
IUPAC Name disodium carbonate
Molecular Weight (g/mol) 105.99
PubChem CID 10340
Equivalence 1 mL = 1 mg CO
ChEBI CHEBI:29377
Grade Laboratory
Packaging Natural Poly Bottle
pH 11 to 11.5
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.0454Normal
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.