Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, 2.00 Normal, Reagents

Catalog No. 01333776
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Catalog No. Quantity
01-333-776 500 mL
01-333-778 1 L
01-333-777 4 L
3 options

Catalog No. 01-333-776

Supplier: Reagents Holdings Llc CS122200500A

Only null left
Add to Cart
Add to Cart

Sodium Carbonate, 2.00 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 497-19-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Soda ash, Disodium carbonate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
pH >12
Physical Form Liquid
Quantity 500 mL
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.101
Chemical Name or Material Sodium Carbonate, 2.00 Normal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.