Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium carbonate, 99+%, for analysis, granular, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
5kg
Packaging:
Plastic Bottle
Plastic drum
497-19-8
CNa2O3
105.99
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
497-19-8
CNa2O3
105.99
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
Biological buffer
Phosphate 10ppm max
Chloride 20ppm max
Packaging Plastic Bottle
Melting Point 851.0°C
Loss on Drying 0.5% max. (285°C)
Color White
Quantity 2.5kg
Boiling Point 1600.0°C
Assay Percent Range 99+%
Linear Formula Na2CO3
Fieser 07,332
Merck Index 15, 8731
Solubility Information Solubility in water: 22 g/100mL (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
Formula Weight 105.99
Physical Form Fine Granular Powder
Percent Purity 99+%
Grade Analysis
Chemical Name or Material Sodium carbonate
Show More Show Less

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed or concerned: Get med

GHS Signal Word: Warning

EINECSNumber : 207-838-8