Learn More
Sodium carbonate, ACS primary standard, 99.95-100.05% (dried basis)
Description
Sodium carbonate is commonly used as an inorganic base and a buffer component in several synthetic organic transformations in both chemical industries and research laboratories. Often used as an electrolyte, sodium carbonate acts as a very good conductor in the process of electrolysis. Apart from being used as a common chemical/biological/biotechnological laboratory reagent, it is also used as a water softener in laundering, processing wood pulp, cleaners, and making glass. It can be used for standardization of acids, and qualitative testing of cations. In dyeing with fiber-reactive dyes, sodium carbonate is used to ensure proper chemical bonding of the dye with cellulose fibers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Granules |
Quantity | 100 g |
Assay Percent Range | 99.95 to 100.05% |
Beilstein | 4154566 |
Solubility Information | Soluble in water. Slightly soluble in aq. alcohol. Insoluble in carbon disulphide,acetone,and alkyl acetates. |
IUPAC Name | disodium carbonate |
Formula Weight | 105.99 |
Grade | ACS Reagent |
Refractive Index | 1.535 |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
missing translation for 'healthHazard1' | H319 |
missing translation for 'healthHazard3' | P234-P264b-P280i-P305+P351+P338-P390 |
EINECSNumber | 207-838-8 |
RTECSNumber | VZ4050000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.