Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium carbonate, anhydrous, Puratronic™, 99.997% (metals basis)

Catalog No. p-4412863
Click to view available options
Quantity:
5 g
25 g
100 g
500 g

CAS: 497-19-8 | CNa2O3 | 105.99 g/mol

Sodium carbonate is commonly used as an inorganic base and a buffer component in several synthetic organic transformations in both chemical industries and research laboratories. Often used as an electrolyte, sodium carbonate acts as a very good conductor in the process of electrolysis. Apart from being used as a common chemical/biological/biotechnological laboratory reagent, it is also used as a water softener in laundering, processing wood pulp, cleaners, and making glass. It can be used for standardization of acids, and qualitative testing of cations. In dyeing with fiber-reactive dyes, sodium carbonate is used to ensure proper chemical bonding of the dye with cellulose fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.997% (metals basis)
Beilstein 4154566
Solubility Information Soluble in water. Slightly soluble in aq. alcohol. Insoluble in carbon disulphide,acetone,and alkyl acetates.
IUPAC Name disodium carbonate
Formula Weight 105.99
Odor Odorless
Refractive Index 1.535
Sensitivity Hygroscopic
Appearance White powder
Density 2.53 g/cm3 at 20°C
Chemical Name or Material Sodium carbonate, anhydrous
Show More Show Less
Hazard Category H319
Precautionary Statement P234-P264b-P280i-P305+P351+P338-P390
EINECSNumber 207-838-8
RTECSNumber VZ4050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.