Sodium carbonate, anhydrous, ACS, 99.5% min. , Alfa Aesar™

Manufacturer: Alfa Aesar 1155218

Quantity 50g

 View more versions of this product

Catalog No. AA1155218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 497-19-8
    Physical Form Granular
    Assay Percent Range 99.5% min.
    Molecular Formula Na2CO3
    Formula Weight 105.99
    MDL Number MFCD00003494
    Chemical Name or Material Sodium carbonate, anhydrous, For AAS analysis
    Grade ACS Grade

    Specifications

    Formula Weight 105.99
    Quantity 50g
    Color White
    Density 2.53g/cm3 at 20°C
    EINECS Number 207-838-8
    Physical Form Granular
    Odor Odorless
    Assay Percent Range 99.5% min.
    Molecular Formula Na2CO3
    Appearance White powder
    MDL Number MFCD00003494
    CAS 497-19-8
    Health Hazard 1 H319: Causes serious eye irritation.
    Chemical Name or Material Sodium carbonate, anhydrous, For AAS analysis
    Grade ACS Grade

    H319: Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit