Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, Anhydrous, Purified (Soda Ash), Reagents

Catalog No. 01337045
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
50 lb.
100 lb.

Sodium Carbonate, Anhydrous, Purified (Soda Ash), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym Soda ash, Disodium carbonate
IUPAC Name disodium carbonate
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Melting Point 851°C
Color Colorless/White
Physical Form Solid
Quantity 100 lb.
Grade Pure
Packaging Fiber Drum
Specific Gravity 2.53
Chemical Name or Material Sodium Carbonate, Anhydrous, Purified (Soda Ash)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.