Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium carbonate, Concentrate Na2CO3 72 mM in Water, IC eluent, Concentrate (20x) for Metrosep A Supp 7, MilliporeSigma™ Supelco™

Catalog No. p-7187741
Click to view available options
Quantity:
1 L
2.5 L

This certified eluent concentrate for ion chromatography is traceable by potentiometric titration to NIST Standard Reference Material. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.

TRUSTED_SUSTAINABILITY

Specifications

CAS 497-19-8
Quantity 1 L
Linear Formula Na2 CO3
MDL Number MFCD00003494
Synonym Sodium carbonate solution
Formula Weight 105.99
Concentration or Composition (by Analyte or Components) 72 mM Na2 CO3 aq. soln.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.