Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium carbonate monohydrate, 99.6%, ACS reagent

Catalog No. p-3735137
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle

CAS: 5968-11-6 | CNa2O3·H2O | 124.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5968-11-6
Molecular Formula CNa2O3·H2O
Molecular Weight (g/mol) 124.01
MDL Number MFCD00149177
InChI Key MQRJBSHKWOFOGF-UHFFFAOYSA-L
Synonym sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
PubChem CID 2735133
IUPAC Name disodium;carbonate;hydrate
SMILES C(=O)([O-])[O-].O.[Na+].[Na+]

Specifications

Melting Point 851.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 99.6%
Linear Formula Na2CO3·H2O
Fieser 07,322
Merck Index 15, 8731
Solubility Information Solubility in water: 210g/L. Other solubilities: soluble in 1.8 parts boiling water, in 7 parts glyceral, insoluble in alcohol
Formula Weight 124.01
Percent Purity 99.6%
Grade ACS Reagent
Loss on Drying 13.0% to 15.0% (150°C)
Packaging Plastic bottle
Chemical Name or Material Sodium carbonate monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.