Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sodium caseinate, pract.

Catalog No. AC419495000
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle

Sodium caseinate, CAS # 9005-46-3, is the salt of the phosphoprotein class found naturally in milk and cheese. It contains all nine essential amino acids and may be used as a molecular reagent, or in cell culture research on growth and differentiation. | CAS: 9005-46-3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Sodium caseinate is the salt of the phosphoprotein class known as caseins
  • Caseins are found in animal tissues, especially milk, and they contain all nine essential amino acids

Applications

  • Casein can be used in protease enzyme activity studies, in vitro experiments on cell growth and differentiation, or as a blocking solution for western blot
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-46-3
MDL Number MFCD00130736

Specifications

Color White to Cream
pH 6 to 7.5 (5% soln.)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500 g
Percent Purity ≥92% (protein) (on dry substance)
Physical Form Powder
Chemical Name or Material Sodium caseinate, pract.
RTECSNumber FI3540000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.