Learn More
Invitrogen™ Sodium Channel Pan Polyclonal Antibody

Description
The antibody detects endogenous levels of total sodium channel protein.
Specifications
Specifications
| Antigen | Sodium Channel Pan |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SCN1A |
| Gene Accession No. | A2APX8, O08562, O88420, O88457, P04774, P04775, P08104, P15389, P15390, P35498, P35499, Q14524, Q15858, Q62205, Q62968, Q6QIY3, Q99250, Q9ER60, Q9JJV9, Q9NY46, Q9R053, Q9UI33, Q9UQD0, Q9WTU3, Q9Y5Y9 |
| Gene Alias | 3-Feb; 6430408L10; A230052E19Rik; AI853486; alternative product; ataxia 3; B230332M13; BFIC3; BFIS3; BFNIS; brain III voltage-gated sodium channel; C630029C19Rik; cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit; CDCD2; CERIII; CIAT; CMD1E; CMPD2; CMS16; CTC-264K15.6; dmu; EIEE11; EIEE13; EIEE6; ETHA; FEB3; FEB3A; FEB3B; FEPS2; FEPS3; FHM3; GEFSP2; GEFSP7; Gm1000; HB1; HB2; HBA; HBBD; HBSC II; HBSCI; HBSCII; HH1; hNa6/Scn8a voltage-gated sodium channel; hNaN; hNE-Na; HOKPP2; hPN3; HSAN2D; HSAN7; HYKPP; HYPP; ICCD; IVF; KIAA1356; Kiaa4197; LQT3; med; mH1; mH2; microI; mKIAA4197; mnd2; mnd-2; Mu-1; Na(v)1.2; Na(V)1.4; Na(V)1.8; Na+ channel; NAC1; NAC1A; NAC2; NAC3; naCh6; NachII; NaN; NaT; Nav1.1; Nav1.2; Nav1.3; Nav1.4; Nav1.5; Nav1.5c; Nav1.6; Nav1.7; Nav1.8; NaV1.9; Nbna1; NCHVS; NENA; NE-NA; Neuroendocrine sodium channel; nmf2; nmf335; nmf58; NSS2; nur14; Peripheral nerve protein type 4; peripheral nerve sodium channel 3; Peripheral nerve sodium channel 5; peripheral sodium channel 1; PFHB1; PN1; PN3; PN4; PN5; RII/RIIA; RIIA sodium channel protein; RNSCPIIR; SCAL; SCIII; SCN; SCN1; SCN10A; SCN11A; SCN12A; SCN1A; SCN2A; SCN2A1; SCN2A2; SCN3A; SCN4A; Scn5a; SCN8A; SCN9A; ScpII; seal; Sensory neuron sodium channel; sensory neuron sodium channel 2; SFNP; skeletal muscle sodium channel alpha subunit; skeletal muscle voltage-dependent sodium channel type IV alpha subunit; skM1; SkM2; SMEI; SNS; Sns2; SNS-2; sodium channel 25; sodium channel 6; sodium channel alpha-subunit; sodium channel protein brain I subunit alpha; Sodium channel protein brain II subunit alpha; sodium channel protein brain III subunit alpha; Sodium channel protein cardiac muscle subunit alpha; sodium channel protein skeletal muscle subunit alpha; Sodium channel protein type 1 subunit alpha; sodium channel protein type 10 subunit alpha; Sodium channel protein type 11 subunit alpha; sodium channel protein type 2 subunit alpha; Sodium channel protein type 3 subunit alpha; sodium channel protein type 4 subunit alpha; Sodium channel protein type 5 subunit alpha; sodium channel protein type 8 subunit alpha; Sodium channel protein type 9 subunit alpha; sodium channel protein type I subunit alpha; Sodium channel protein type II subunit alpha; sodium channel protein type III subunit alpha; Sodium channel protein type IV subunit alpha; sodium channel protein type IX subunit alpha; Sodium channel protein type V subunit alpha; sodium channel protein type VIII subunit alpha; Sodium channel protein type X subunit alpha; sodium channel protein type XI subunit alpha; sodium channel protein, brain I alpha subunit; sodium channel protein, brain I subunit alpha; sodium channel protein, brain II subunit alpha; sodium channel protein, brain III subunit alpha; sodium channel protein, brain type 2 alpha subunit; sodium channel type X alpha polypeptide; sodium channel voltage gated type 1 alpha subunit; sodium channel voltage-gated type 11 alpha polypeptide; sodium channel voltage-gated type 4 alpha polypeptide; sodium channel voltage-gated type I alpha polypeptide; sodium channel voltage-gated type II alpha polypeptide; Sodium channel voltage-gated type IV alpha polypeptide; sodium channel voltage-gated type V alpha polypeptide; sodium channel voltage-gated type VIII alpha polypeptide; sodium channel voltage-gated type X alpha polypeptide; sodium channel voltage-gated type XI alpha polypeptide; sodium channel, voltage gated, type I alpha subunit; sodium channel, voltage gated, type II alpha subunit; sodium channel, voltage gated, type III alpha subunit; sodium channel, voltage gated, type IV alpha subunit; sodium channel, voltage gated, type IX alpha subunit; sodium channel, voltage gated, type V alpha subunit; sodium channel, voltage gated, type VIII alpha subunit; sodium channel, voltage gated, type VIII, alpha subunit; sodium channel, voltage gated, type X alpha subunit; sodium channel, voltage gated, type XI alpha subunit |
| Gene Symbols | SCN10A, Scn11a, SCN1A, SCN2A, SCN3A, Scn4a, Scn5a, Scn8a, SCN9A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.