Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCN11A Polyclonal Antibody

Sodium Channel Pan Polyclonal Antibody
Supplier: Thermo Scientific PA5104156
Description
Sodium Channel Pan Polyclonal Antibody for Western Blot

Specifications
SCN11A | |
Polyclonal | |
Unconjugated | |
SCN11A | |
FEPS3; hNaN; HSAN7; Nan; NaT; NaV1.9; NSS2; Peripheral nerve sodium channel 5; PN5; Scn11a; SCN12A; sensory neuron sodium channel 2; SNS2; SNS-2; Sodium channel protein type 11 subunit alpha; sodium channel protein type XI subunit alpha; sodium channel voltage-gated type 11 alpha polypeptide; sodium channel voltage-gated type XI alpha polypeptide; sodium channel, voltage gated, type XI alpha subunit; sodium channel, voltage-gated, type 11, alpha polypeptide; sodium channel, voltage-gated, type XI, alpha; sodium channel, voltage-gated, type XI, alpha polypeptide; sodium channel, voltage-gated, type XI, alpha subunit; sodium channel, voltage-gated, type XII, alpha polypeptide; sodium channel, voltage-gated, type11, alpha polypeptide; sodium voltage-gated channel alpha subunit 11; voltage-gated sodium channel NAV1.9b; voltage-gated sodium channel subunit alpha Nav1.9 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11280 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9UI33 | |
SCN11A | |
A synthesized peptide derived from human SCN11A(Accession Q9UI33), corresponding to amino acid residues E473-L523. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction