Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium chloranilate trihydrate, 95%, Thermo Scientific™

Catalog No. ac419500250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 36275-66-8
Molecular Formula C6Cl2Na2O4·3H2O
Molecular Weight (g/mol) 307
MDL Number MFCD00150173
InChI Key FITANBHHCLWRKG-UHFFFAOYSA-L
Synonym chloranilic acid sodium salt, chloranilic acid disodium salt, disodium chloranilate, chloroanilic acid sodium salt, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy-, disodium salt, sodium chloranilate, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy-, sodium salt 1:2, disodium 2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-diolate, chloranilic acid sodium salt hydrate, disodium 2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-bis olate
PubChem CID 118938
IUPAC Name disodium;2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-diolate
SMILES C1(=C(C(=O)C(=C(C1=O)Cl)[O-])Cl)[O-].[Na+].[Na+]

Specifications

Color Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 25 g
Formula Weight 307
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Sodium chloranilate trihydrate, 95%
EINECSNumber 252-947-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.