Learn More
Sodium chlorate, 99% min
Description
Sodium chlorate is used to prepare chlorine dioxide in the pulp and paper industry. It is utilized for bleaching the wood pulp in order to make environmentally friendly and quality white paper products. It is also used to produce other perchlorate compounds in the industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 248°C |
Color | Colorless |
Physical Form | Granules |
Quantity | 1 kg |
Assay Percent Range | 99% min. |
Solubility Information | Soluble in water,glycerol,hydrazine,acetone and methanol. Slightly soluble in ethanol and ammonia. |
Formula Weight | 106.45 |
Odor | Odorless |
Appearance | Colorless granules |
Density | 2.5 g/cm3 |
Show More |
Safety and Handling
Hazard Category | H271-H302-H373 |
Precautionary Statement | P210-P220-P221-P260-P264b-P270-P280-P283-P301+P312-P306+P360-P314-P330-P370+P378q-P371+P380+P375-P501c |
DOTInformation | Hazard Class: 5.1, Packing Group: II |
EINECSNumber | 231-887-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.