Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium chlorate, ACS, 99.0% min

Catalog No. p-4407872
Click to view available options
Quantity:
100 g
500 g
2 kg

Maximum level of impurities: Insoluble matter 0.005%, BrO3 0.015%, Cl 0.005%, SO4 0.001%, Heavy metals (as Pb) 0.001%, Fe 5ppm, Ca 0.002%, Mg 0.002%, K 0.01% | CAS: 7775-09-9 | ClNaO3 | 106.44 g/mol

Sodium chlorate is used to prepare chlorine dioxide in the pulp and paper industry. It is utilized for bleaching the wood pulp in order to make environmentally friendly and quality white paper products. It is also used to produce other perchlorate compounds in the industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-09-9
Molecular Formula ClNaO3
Molecular Weight (g/mol) 106.44
MDL Number MFCD00003479
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
Synonym sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
PubChem CID 516902
ChEBI CHEBI:65242
SMILES [Na+].[O-][Cl](=O)=O

Specifications

Melting Point 248°C
Color Colorless
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99% min.
Solubility Information Soluble in water,glycerol,hydrazine,acetone and methanol. Slightly soluble in ethanol and ammonia.
IUPAC Name sodium chlorate
Formula Weight 106.45
Grade ACS
Odor Odorless
Density 2.5 g/cm3 at 20°C
Chemical Name or Material Sodium chlorate, For analysis ACS
Show More Show Less
Hazard Category H271-H302-H373
Precautionary Statement P210-P220-P221-P260-P264b-P270-P280-P283-P301+P312-P306+P360-P314-P330-P370+P378q-P371+P380+P375-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 231-887-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.