Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, 0.1 Normal Ionic Strength Adjustor, Reagents

Catalog No. 01333963
Change view
Click to view available options
Quantity:
500 mL
1 L
Catalog No. Quantity
01-333-963 1 L
01-333-962 500 mL
2 options

Catalog No. 01-333-963

Supplier: Reagents Holdings Llc CS0111001A

Only null left
Add to Cart
Add to Cart

Sodium Chloride, 0.1 Normal Ionic Strength Adjustor, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-14-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Table salt solution
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
pH 6.7 to 7.3
Physical Form Liquid
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0032
Chemical Name or Material Sodium Chloride, 0.1 Normal Ionic Strength Adjustor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.