Learn More
Sodium chloride, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Sodium chloride is widely used as a food additive, and a preservative. It is employed in the Chloralkali process for producing chlorine and sodium hydroxide, in the Solvay process for producing sodium carbonate and calcium chloride, in the Mannheim process and in the Hargreaves process for producing sodium sulfate and hydrochloric acid. It is used in the manufacture of sodium chlorate, water softening and de-icing of roadways. Further it is used in several industries like pharmaceutical, textile, dyeing, rubber food and agriculture. It is used in the processing of steel, copper, vanadium, and other metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1413°C |
Melting Point | 801°C |
Color | White |
Physical Form | Crystals |
Quantity | 50 g |
Assay Percent Range | 99.99% (metals basis) |
Solubility Information | Freely soluble in water. Slightly soluble in alcohol and ammonia. |
IUPAC Name | sodium;chloride |
Formula Weight | 58.44 |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H303: May be harmful if swallowed. |
EINECSNumber | 231-598-3 |
RTECSNumber | VZ4725000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.