Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Chloride, ACS Reagent, Reagents
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
12 kg
50 kg
350 lb.
Specifications
Specifications
| Boiling Point | 1461°C |
| Melting Point | 801°C |
| Color | Colorless/White |
| pH | 6.7 to 7.3 |
| Physical Form | Solid |
| Quantity | 350 lb. |
| Grade | ACS |
| Packaging | Fiber Drum |
| Specific Gravity | 2.17 |
| Chemical Name or Material | Sodium Chloride, ACS Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction