Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, Saturated, Ricca Chemical
SDP

Catalog No. r7227600500
Click to view available options
Quantity:
500 mL

Chemical Identifiers

CAS 7732-18-5, 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

pH 6.7 to 7.3
Physical Form Solution
Quantity 500 mL
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Odorless
Packaging Natural Poly Bottle
Appearance Clear colorless liquid
Chemical Name or Material Sodium Chloride
Recommended Storage General
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.