Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Chloride, Crystal, ACS, 99%, Spectrum™ Chemical

Click to view available options
Quantity:
500 g
1 kg
2.5 kg
12 kg
45 kg
350 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
Specifications
Specifications
pH | 5.0 to 9.0 |
CAS Max % | 1 |
Quantity | 2.5 kg |
Percent Purity | 99% |
Grade | ACS |
Identification | To Pass Test |
Packaging | Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction