Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride (Crystalline/Certified ACS), Fisher Chemical PROMO

Catalog No. S2713FB25 Shop All Fisherbrand Products
Click to view available options
:
3 kg

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44 g/mol
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, Sodium chloride, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name Sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461°C
Melting Point 801°C
Color White
pH 5.0 to 8.0
Physical Form Solid
Quantity 3 kg
Assay Percent Range ≥99.0%
Solubility Information Solubility in Water: Partially Soluble
Formula Weight 58.44 amu
Percent Purity ≥99.0%
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Specific Gravity 2.165
Vapor Pressure 0 mmHg at 440°C
Density 2.165 g/cm3
Chemical Name or Material Sodium chloride
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement

Emergency Overview: May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1

Flammability:1

Instability:0

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.