Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium chloride, extra pure

Catalog No. AC194090250
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle

Catalog No. AC194090250

Supplier: Thermo Scientific Chemicals 194090250

Only null left
Add to Cart

CAS: 7647-14-5 | ClNa | 58.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461.0°C
Melting Point 801.0°C
Color White
pH 5.0 to 8.0 (5%, H<2O, at 20°C)
Physical Form Crystalline Powder
Quantity 25 kg
Linear Formula NaCl
Fieser 13,280
Merck Index 15, 8734
Solubility Information Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol (1g/10mL), ammonia, slightly soluble in alcohol, nearly insoluble in hydrochloric acid
IUPAC Name sodium;chloride
Formula Weight 58.44
Percent Purity ≥99.0%
Grade Extra Pure
Packaging Fibre drum
Chemical Name or Material Sodium chloride
Show More Show Less
EINECSNumber 231-598-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.