Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Sodium Chloride, Lab Grade
Shop All Fisherbrand Products

Click to view available options
Quantity:
500 g
2.5 kg
3 kg
12 kg
Description
This product is for educational use - no C of A is available.
Specifications
Specifications
Color | White |
Quantity | 3 kg |
Assay Percent Range | >95% w/v |
Linear Formula | NaCl |
Formula Weight | 58.44 |
Grade | Laboratory |
Density | 2.165g/mL |
Chemical Name or Material | Sodium Chloride |
Safety and Handling

ShelfLife | 5 years |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction